Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Esculin hydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
531-75-9
C15H16O9·xH2O
340.29
MFCD00149492
XHCADAYNFIFUHF-TVKJYDDYSA-N
esculin,aesculin,esculoside,--esculin,polychrome,6,7-dihydroxycoumarin-6-o-glucoside,aesculinum,esculine,esculetin 6-o-glucoside,6,7-dihydroxycoumarin 6-glucoside
5281417
CHEBI:4853
7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O
531-75-9
C15H16O9·xH2O
340.29
MFCD00149492
XHCADAYNFIFUHF-TVKJYDDYSA-N
esculin,aesculin,esculoside,--esculin,polychrome,6,7-dihydroxycoumarin-6-o-glucoside,aesculinum,esculine,esculetin 6-o-glucoside,6,7-dihydroxycoumarin 6-glucoside
5281417
CHEBI:4853
7-hydroxy-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
C1=CC(=O)OC2=CC(=C(C=C21)OC3C(C(C(C(O3)CO)O)O)O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 203.0°C to 205.0°C
Specific Rotation Condition - 86.00 (20.00°C c=2,dioxane/water1:1)
Color Undesignated
Quantity 50g
Sulfated Ash 0.1% max.
Water 8% max. (K.F.)
Specific Rotation − 86.00
Assay Percent Range 97%
Merck Index 15, 3753
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in hot water,soluble in hot alcohol,methanol,pyridine,,ethyl acetate and acetic acid
Formula Weight 340.29
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Esculin hydrate
Show More Show Less