Learn More
White Knight™ ESD Knit Waist Length Lab Coat
Offering superior protection against static build up and electrostatic discharge. Our ESD lab coats are the obvious choice for your industrial environment needs.
$62.50 - $101.00
Specifications
Certifications/Compliance Qualifies as a groundable static control garment
Closure Type Snaps
Collar Type Lapel
Color Blue
Cuff Style Stud-fastened
View More Specs
Products 10
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-111-338
View Documents
White Knight™
12L154FCESDK01XSBLU
X-Small
Each for $62.50
Add to cart
18-111-339
View Documents
White Knight™
12L154FCESDK01SBLU
Small
Each for $62.50
Add to cart
18-111-340
View Documents
White Knight™
12L154FCESDK01MBLU
Medium
Each for $62.50
Add to cart
18-111-341
View Documents
White Knight™
12L154FCESDK01LBLU
Large
Each for $62.50
Add to cart
18-111-342
View Documents
White Knight™
12L154FCESDK01XLBLU
X-Large
Each for $62.50
Add to cart
18-111-343
View Documents
White Knight™
12L154FCESDK012XLBLU
2X-Large
Each for $68.75
Add to cart
18-111-344
View Documents
White Knight™
12L154FCESDK013XLBLU
3X-Large
Each for $76.00
Add to cart
18-111-345
View Documents
White Knight™
12L154FCESDK014XLBLU
4X-Large
Each for $83.00
Add to cart
18-111-346
View Documents
White Knight™
12L154FCESDK015XLBLU
5X-Large
Each for $92.00
Add to cart
18-111-347
View Documents
White Knight™
12L154FCESDK016XLBLU
6X-Large
Each for $101.00
Add to cart
Description
Description
  • Reinforced, adjustment cuff snaps
  • Reinforced snap front closure
  • Grounding snaps on pockets and under arms
  • ESD compliance indicator label
  • Reusable ESD Knit fabric
  • Qualifies as a groundable, static control garment
Specifications
Specifications
Qualifies as a groundable static control garment
Lapel
Stud-fastened
Unisex
ESD Knit Polyester
Pre-serged/Felled
Snaps
Blue
No
Yes
3
Non-sterile