Learn More
ESF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 234961AP
Catalog No. 50-173-4084
Add to cart
Description
Description
Production of cholesterol by reduction of C7-C8 double bond of 7-dehydrocholesterol (7-DHC).
Specifications
Specifications
ESF1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3V1V3, Q9H501
ABT1 associated protein, ABTAP, C20orf6, ESF1, ESF1 homolog, HDCMC28P
Rabbit
IgG
150 μL
-20°C
Primary
51575, 66580
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
ESF1
Liquid
ESF1
ESF1 Fusion Protein Ag19037
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse