Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Esmolol hydrochloride

Click to view available options
Quantity:
5 g
25 g
81161-17-3
C16H26ClNO4
331.84
GEKNCWBANDDJJL-UHFFFAOYNA-N
methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate hydrochloride
Cl.COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1
81161-17-3
C16H26ClNO4
331.84
GEKNCWBANDDJJL-UHFFFAOYNA-N
methyl 3-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)propanoate hydrochloride
Cl.COC(=O)CCC1=CC=C(OCC(O)CNC(C)C)C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 86.0°C to 92.0°C
Quantity 5 g
Formula Weight 331.83
Chemical Name or Material Esmolol hydrochloride

GHS H Statement Causes serious eye irritation.

GHS P Statement Wear eye protection/face protection.

Wash face, hands and any exposed skin thoroughly after handling.

If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

RUO – Research Use Only