Learn More
Abnova™ ESR2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00002100T03
Catalog No. 89-943-629
Add to cart
Specifications
Specifications
Western Blot
293T
BC024181.2
pCMV-ESR2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
ESR2 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 35.9kDa
Transient overexpression cell lysate was tested with anti-ESR2 antibody (Mfr. No. H00002100-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)