Learn More
ESS™ Profile NVG™ Goggle
Compact military/tactical goggle created with direction from elite U.S. Special Forces groups
Manufacturer:  ESS™ 7400124
 View more versions of this product
Catalog No. 19-121-824
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Extra-thick 2.8mm lenses provide 100% UVA/UVB protection and distortion-free optics
  • Low-profile frame offers dust filtration, clear weapon sighting and field of view, impact protection, and anti-fog performance
  • Fully filtered, high airflow vents surround the entire lens to minimize fogging while repelling particles and smoke
  • Soft, conforming frame has comfortable, sealed fit on all face shapes, and smooth interface is easy to clean—won't become contaminated by face paint
  • Universal helmet and night vision compatibility
Compliance
Meets rigorous MIL-V-43511C .22 caliber ballistic impact test and all ANSI Z87.1 requirements.
Specifications
Specifications
Tactical Goggle
Scratch-resistant, Anti-fog, Anti-UV
Tactical Black
Filtered, high airflow vents
Clear
MIL-V-43511C .22 caliber ballistic impact test and all ANSI Z87.1
99.90%
Antifog; Scratch-Resistant