Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Essential iPSC Factor Antibody Pack

For Research applications

Manufacturer:  Novus Biologicals™ NBP142824

Catalog No. NBP142824


Add to cart

Specifications

Specifications

Essential iPSC Factor
Unconjugated
Affinity Purified
NB100-588-A synthetic peptide which maps to a region between residues 250 and the C-terminus (residue 305) of mouse Nanog homeobox using the numbering given in TrEMBL entry Q80Z64 (GeneID 71950). NB100-2379-A synthetic peptide made to an internal portion
6 Vials
Store at 4C. Do not freeze.
Bovine, Chicken, Human, Mouse, Porcine, Primate, Sheep
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Rabbit
Immunogen affinity purified
Stem Cell Markers
1.NB100-2379: OCT4 Antibody
2.NB100-588: Nanog Antibody
3.NB110-37235: SOX2 Antibody
4.NB600-302: c-Myc Antibody (9E10)
5.NBP1-28865: LIN-28A Antibody (6D1F9)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit