Learn More
Crystal Chem IncSupplier Diversity Partner Estradiol ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80962
The Estradiol ELISA is a highly sensitive assay that is used to quantify levels of human estradiol in serum. Highlights: Kit uses sample size of only 25 µL. High sensitivity of 10 pg/mL. Wide dynamic range of 10 - 1000 pg/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7868