Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estradiol, Hemihydrate, Soy Base, Micronized, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
35380-71-3
C36H50O5
562.79
ZVVGLAMWAQMPDR-WVEWYJOQSA-N
bis((1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol) hydrate
O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O
35380-71-3
C36H50O5
562.79
ZVVGLAMWAQMPDR-WVEWYJOQSA-N
bis((1S,3aS,3bR,9bS,11aS)-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1,7-diol) hydrate
O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O.C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CC[C@@H]2O

E2918, 35380-71-3, C18H24O21/2H2O

Spectrum™ Estradiol, Hemihydrate, Soy Base, Micronized, USP
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation +76° to +83°
Melting Range 173°-179°C
CAS Max % 1
Percent Purity 97 to 103%