Learn More
Estradiol Recombinant Rabbit Monoclonal Antibody (RM343), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA527910
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Estradiol Recombinant Monoclonal antibody specifically detects Estradiol in Chemical samples. It is validated for ELISA.Specifications
Estradiol | |
Recombinant Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
Rabbit | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG |
ELISA | |
RM343 | |
Unconjugated | |
17-Beta-Estradiol; 17-beta-HSD | |
Estradiol BSA Conjugate | |
100 μL | |
Primary | |
-20°C | |
Liquid | |
Chemical |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.