Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrogen Receptor beta Mouse anti-Human, Clone: MC10, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14933682

Encompass

 View more versions of this product

Catalog No. 50-112-2779


Add to cart

Description

Description

Description: The MC10 monoclonal antibody recognizes all forms of human estrogen receptor (ER) beta. ER beta is a member of a superfamily of nuclear receptors, which are ligand-dependent transcription factors. ER beta like ER alpha, becomes activated by binding the hormone estrogen, causing a dissociation from chaperone proteins and subsequent dimerization with either ER beta or ER alpha. Unlike ER alpha, the result of ER beta signaling is thought to be anti-proliferative and proapoptotic. The MC10 clone recognizes full length ER beta as well as each of its splice variants. Applications Reported: This MC10 antibody has been reported for use in immunoprecipitation, western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, ELISA, and immunocytochemistry. Applications Tested: This MC10 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using either low or high pH antigen retrieval and can be used at less than or equal to 30 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.

Estrogen Receptor beta (ER beta, NR3A2) protein is approximately 55kD, though multiple isoforms ranging from 36-59kD have been described. Human ER beta was initially cloned and characterized from testis. Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Estrogen receptor beta (ER beta) binds estrogens with an affinity similar to that of estrogen receptor alpha. Some ER beta isoforms domitly inhibit the activity of estrogen receptor alpha in reproductive tissues. ER beta is found widely in many tissues throughout the body and can act as a potent tumor suppressor, playing a crucial role in many cancer types such as prostate cancer, autism spectrum disorder, medullary thyroid carcinoma, De Quervain Disease and risk of cardiovascular disease.
Specifications

Specifications

Estrogen Receptor beta
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q92731
ESR2
Affinity chromatography
RUO
2100
4° C
Liquid
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
MC10
Unconjugated
ESR2
ER Beta, ER, ESR2, ESR-Beta
Mouse
100 μg
Primary
Human
Antibody
IgG2a κ
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.