Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Estrogen Receptor (SP1), Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Estrogen Receptor (SP1), Rabbit Monoclonal Antibody.

$469.00 - $2510.00

Specifications

Antigen Estrogen Receptor (SP1)
Clone SP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Rabbit
View More Specs

Products 5
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
RM9101R7
View Documents
Thermo Scientific™
RM9101R7
Ready-to-use for IHC (UVLP) 7mL
Each for $1,077.00
Add to cart
 
RM9101RQ
View Documents
Thermo Scientific™
RM9101RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $1,370.00
Add to cart
 
RM9101S
View Documents
Thermo Scientific™
RM9101S
Culture Supernatant with Azide 1mL
Each for $2,510.00
Add to cart
 
RM9101S0
View Documents
Thermo Scientific™
RM9101S0
Culture Supernatant with Azide 100μL
Each for $469.00
Add to cart
 
RM9101S1
View Documents
Thermo Scientific™
RM9101S1
Culture Supernatant with Azide 500μL
Each for $1,755.00
Add to cart
 
Description

Description

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Thermo Scientific™ Estrogen Receptor (SP1), Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Recognizes a protein of 67 kDa, which is identified as estrogen receptor (ER) alpha. The ER gene consists of more than 140kb of genomic DNA divided into 8 exons, and is translated into a protein with six functionally discrete domains, labeled A through F. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Host Species: Rabbit

Clone: SP1

Isotype: IgG

Species Reactivity: Human, Mouse and Hamster. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide derived from C-terminal of human estrogen receptor

Molecular Weight: 67 kDa

Positive Control: Breast CA, MCF-7 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence (Not verified)
  • Flow Cytometery (Not verified)

Specifications

Specifications

Estrogen Receptor (SP1)
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Rabbit
Steroids Hormones and Growth Factors
Synthetic peptide derived from C-terminal of human estrogen receptor
SP1
Monoclonal
IVD
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit