Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Estrone, Plant Based, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
53-16-7
C18H22O2
270.37
DNXHEGUUPJUMQT-CBZIJGRNSA-N
(3aS,3bR,9bS,11aS)-7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CCC2=O
53-16-7
C18H22O2
270.37
DNXHEGUUPJUMQT-CBZIJGRNSA-N
(3aS,3bR,9bS,11aS)-7-hydroxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-one
C[C@]12CC[C@H]3[C@@H](CCC4=CC(O)=CC=C34)[C@@H]1CCC2=O

E1439, 53-16-7, C18H22O2

Spectrum™ Estrone, Plant Based, USP is a sex hormone that is used in hormone replacement therapy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.005
Loss on Drying 0.005
Specific Rotation +158° to +165°
Quantity 5 g
CAS Max % 1
Percent Purity 97 to 103%