Learn More
ETFA Antibody (OTI5C4), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270693H
Catalog No. NBP270693H
Add to Cart
Description
Description
ETFA Monoclonal specifically detects ETFA in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
ETFA
Monoclonal
HRP
PBS with No Preservative
ETFA
Full length recombinant protein of human ETFA (NP_000117) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot
OTI5C4
Western Blot
alpha-ETF, electron transfer flavoprotein alpha-subunit, Electron transfer flavoprotein subunit alpha, mitochondrial, electron-transfer-flavoprotein, alpha polypeptide, EMA, GA2, glutaric aciduria II, MADD
Mouse
Affinity Purified
Core ESC Like Genes, Stem Cell Markers
2108.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions