Learn More
ETFA Mouse anti-Human, Clone: OTI5C4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF808409
Catalog No. 50-166-8834
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ETFA participates in catalyzing the initial step of the mitochondrial fatty acid beta-oxidation. It shuttles electrons between primary flavoprotein dehydrogenases and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. Defects in electron-transfer-flavoprotein have been implicated in type II glutaricaciduria in which multiple acyl-CoA dehydrogenase deficiencies result in large excretion of glutaric, lactic, ethylmalonic, butyric, isobutyric, 2-methyl-butyric, and isovaleric acids. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ETFA
Monoclonal
Unconjugated
ETFA
Lyophilized
ETFA
Full length human recombit protein of human ETFA produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5C4
PBS with 8% trehalose and no preservative; pH 7.3
P13804
EMA, GA2, MADD
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2108