Learn More
Tocris Bioscience™ Ethacrynic acid - d5 Available on GSA/VA Contract for Federal Government customers only.
Deuterated ethacrynic acid
Manufacturer:  Tocris Bioscience™ 4987/1
Catalog No. 49-871-R
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
Deuterated ethacrynic acid. Used as an internal standard for GC- or LC-mass spectrometry.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Ethacrynic acid - d5
>99
1mg
Ethacrynic acid - d5
308.17