Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethambutol, 98%, Alfa Aesar™

Click to view available options
Quantity:
25g
74-55-5
C10H24N2O2
204.314
MFCD00242828
AEUTYOVWOVBAKS-UWVGGRQHSA-N
14052
CHEBI:4877
(2S)-2-[2-[[(2S)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol
CCC(CO)NCCNC(CC)CO
74-55-5
C10H24N2O2
204.314
MFCD00242828
AEUTYOVWOVBAKS-UWVGGRQHSA-N
14052
CHEBI:4877
(2S)-2-[2-[[(2S)-1-hydroxybutan-2-yl]amino]ethylamino]butan-1-ol
CCC(CO)NCCNC(CC)CO

Used in conjunction with at least one other antituberculous drug in the treatment of pulmonary tuberculosis.

Solubility
Soluble in water. Also soluble in chloroform and methylene chloride

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Solubility Information Soluble in water. Also soluble in chloroform and methylene chloride
Formula Weight 204.31
Physical Form Powder
Percent Purity 98%
Boiling Point 345°C
Color White
Melting Point 199°C to 204°C
Quantity 25g
Flash Point >110°C(230°F)
Chemical Name or Material Ethambutol

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EL3640000

storageNote1 : 2° to 8°C