Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethambutol Hydrochloride, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

E1262, 1070-11-7, C10H24N2O2•2HCl

Spectrum™ Ethambutol Hydrochloride, USP is a bacteriostatic antimycobacterial drug used to treat tuberculosis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Loss on Drying 0.005
Quantity 25 kg
Specific Rotation +6.0° to +6.7°
CAS 1070-11-7
CAS Max % 1
Molecular Formula C10H26Cl2N2O2
InChI Key AUAHHJJRFHRVPV-BZDVOYDHSA-N
SMILES [H+].[H+].[Cl-].[Cl-].CC[C@@H](CO)NCCN[C@@H](CC)CO
IUPAC Name dihydrogen (2S)-2-[(2-{[(2S)-1-hydroxybutan-2-yl]amino}ethyl)amino]butan-1-ol dichloride
Molecular Weight (g/mol) 277.23
Percent Purity 98 to 100.5%
Show More Show Less