Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanesulfonic acid, 96%, Thermo Scientific Chemicals

Catalog Number AA1489118
Click to view available options
Quantity:
10 g
50 g
250 g
594-45-6
C2H5O3S
109.12
MFCD00007529
CCIVGXIOQKPBKL-UHFFFAOYSA-M
ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
11668
CHEBI:42465
ethanesulfonic acid
CCS([O-])(=O)=O
594-45-6
C2H5O3S
109.12
MFCD00007529
CCIVGXIOQKPBKL-UHFFFAOYSA-M
ethylsulfonic acid, ethanesulphonic acid, ethane sulfonate, ethane sulfonic acid, ethane sulphonic acid, ethansulfonic acid, ethylsulphonic acid, 1-ethanesulfonic acid, 2-ethanesulfonic acid, ethanesulfonicacid
11668
CHEBI:42465
ethanesulfonic acid
CCS([O-])(=O)=O

Ethanesulfonic acid is employed in the electrolytic reduction of perrhenate solutions. It is also used as a catalyst for alkylation, polymerization and other reactions. Further, it reacts with triethoxymethane to prepare ethanesulfonic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -17°C
Color Colorless to Yellow
Density 1.343 g/mL
Boiling Point 122°C to 123°C (0.01 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.435
Quantity 50 g
UN Number UN2586
Beilstein 1743071
Solubility Information Miscible with water.
Formula Weight 110.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethanesulfonic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID
EINECSNumber 209-843-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit