Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethanesulfonyl chloride, 98+%

Click to view available options
Quantity:
25g
100g
500g
594-44-5
C2H5ClO2S
128.57
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride,ethylsulfochloride,1-ethanesulfonyl chloride,ethanesulphonyl chloride,ethanesulfonylchloride,unii-b56zoc898a,ethylsulfonylchloride,ethansulfonyl chloride,ethansulfonylchlorid,ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(=O)(=O)Cl
594-44-5
C2H5ClO2S
128.57
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride,ethylsulfochloride,1-ethanesulfonyl chloride,ethanesulphonyl chloride,ethanesulfonylchloride,unii-b56zoc898a,ethylsulfonylchloride,ethansulfonyl chloride,ethansulfonylchlorid,ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(=O)(=O)Cl
Density 1.35
Refractive Index 1.454
Sensitivity Moisture sensitive
Melting Point -70°C
Flash Point 83°C (181°F)
Boiling Point 176°C to 178°C
Quantity 25g
Linear Formula CH3CH2SO2Cl
UN Number UN2927
Beilstein 773865
Formula Weight 128.57
Percent Purity ≥98%
Chemical Name or Material Ethanesulfonyl chloride
Show More Show Less

GHS H Statement
H301-H310-H330-H314-H318-H227
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314-H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 209-842-5

RTECSNumber : KI8050000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen