Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethanesulfonyl chloride, 98+%

Click to view available options
Quantity:
25g
100g
500g
594-44-5
C2H5ClO2S
128.57
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride,ethylsulfochloride,1-ethanesulfonyl chloride,ethanesulphonyl chloride,ethanesulfonylchloride,unii-b56zoc898a,ethylsulfonylchloride,ethansulfonyl chloride,ethansulfonylchlorid,ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(=O)(=O)Cl
594-44-5
C2H5ClO2S
128.57
MFCD00007460
FRYHCSODNHYDPU-UHFFFAOYSA-N
ethylsulfonyl chloride,ethylsulfochloride,1-ethanesulfonyl chloride,ethanesulphonyl chloride,ethanesulfonylchloride,unii-b56zoc898a,ethylsulfonylchloride,ethansulfonyl chloride,ethansulfonylchlorid,ethanesulfonylchoride
11667
ethanesulfonyl chloride
CCS(=O)(=O)Cl
Boiling Point 176°C to 178°C
Melting Point -70°C
Flash Point 83°C (181°F)
Quantity 100g
Linear Formula CH3CH2SO2Cl
UN Number UN2927
Beilstein 773865
Formula Weight 128.57
Percent Purity ≥98%
Chemical Name or Material Ethanesulfonyl chloride

GHS H Statement
H301-H310-H330-H314-H318-H227
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : KI8050000

TSCA : TSCA