Ethanol, 140 proof (70%), Decon™ Labs

Manufacturer: Decon Laboratories, Inc.Supplier Diversity Partner 2455M

 View more versions of this product

Catalog No. 04-355-175

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Boiling Point 78.3°C
    Melting Point −114°C
    Quantity 55 gal.
    DOT Information DOT Class: 3, Packaging: II
    Density 7.109 lb./gal.
    Physical Form Liquid
    UN Number UN1170
    Recommended Storage Do not store alcohol above 120°F
    CAS 62.0,7732-18-5 (purified water)
    Chemical Name or Material Ethanol
    Packaging Metal Drum
    Name Note 140 Proof (70%)
    CAS Min % 62.0
    CAS Max % 70.0

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit