Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 140 proof (70%), Tax-Paid, Decon™ Labs

Catalog Number 04355351
Click to view available options
Quantity:
5 gal.
4 x 1 gal.
Packaging:
Plastic Bottle
64-17-5, 7732-18-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
64-17-5, 7732-18-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO

Federal Tax is included in the price

Product 07-678-001: May require a federal tax exempt certificate.

Melting Point -114°C
Density 7.109
Boiling Point 78.3°C
Quantity 5 gal.
Packaging Plastic Bottle
Physical Form Liquid
UN Number UN1170
Chemical Name or Material Ethanol, 140 Proof (70%)
DOTInformation DOT Class: 3, Packaging: II
Recommended Storage Do not store alcohol above 120°F
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit