Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 140 proof (70%), Tax-Paid, Decon™ Labs

Click to view available options
Quantity:
5 gal.
4 x 1 gal.
Packaging:
Plastic Bottle
64-17-5, 7732-18-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO
64-17-5, 7732-18-5
C2H6O
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alkohol, anhydrol, tecsol
702
CHEBI:16236
ethanol
CCO

Federal Tax is included in the price

UN Number UN1170
Physical Form Liquid
Density 7.109 lb./gal.
Packaging Plastic Bottle
Boiling Point 78.3°C
Melting Point -114°C
Quantity 4 x 1 gal.
Chemical Name or Material Ethanol, 140 Proof (70%)

DOTInformation : DOT Class: 3, Packaging: II

Recommended Storage : Do not store alcohol above 120°F