Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanol, 190 proof (95%), Tax-Paid, Decon™ Labs
Click to view available options
Quantity:
0.125 gal.
1 gal.
5 gal.
4 x 1 gal.
55 gal.
Packaging:
Drum
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle with Spigot Closure
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
CAS Min % | 94.9 |
CAS Max % | 96.0 |
UN Number | 1170 |
Physical Form | Liquid |
Density | 6.72g/mL |
Packaging | Plastic Bottle |
Boiling Point | 78.3°C |
Melting Point | -114°C |
Quantity | 0.125 gal. |
Chemical Name or Material | Ethanol, 190 Proof (95%) |
Safety and Handling
DOTInformation : DOT Class: 3, Packaging: II
Recommended Storage : Do not store alcohol above 120°F