Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanol, 190 proof (95%), USP, Decon™ Labs
Click to view available options
Quantity:
5 gal.
4 x 1 gal.
55 gal.
24 x 0.125 gal.
Packaging:
Cubitainer with Spigot
Glass Bottle
Metal Container
N/A
Plastic Bottle
Poly Bottle
Poly Bottle (Closure can be fitted with spigot)
Poly Drum
Description
Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.
Specifications
Melting Point | -113.9°C |
CAS Min % | 99.9 |
CAS Max % | 100.0 |
UN Number | UN1170 |
Grade | BP/EP/FCC/JP/NF/USP |
Density | 6.72 |
Boiling Point | 78.3°C |
Quantity | 24 x 0.125 gal. |
Packaging | Plastic Bottle |
Physical Form | Liquid |
Show More |
Safety and Handling
DOTInformation | DOT Class: 3, Packaging: II |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction