Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 190 proof (95%), USP, Decon™ Labs

May require a federal tax exempt certificate.

Manufacturer:  Decon Laboratories, Inc.Supplier Diversity Partner 2816TPD

 View more versions of this product

Catalog No. 04355453D


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.
Description & Specifications

Specifications

Ethanol
99.9,7732-18-5 (purified water)
100.0
Liquid
Ethyl Alcohol
N/A
−113.9°C
190 Proof (95%)
99.9
BP/EP/FCC/JP/NF/USP
UN1170
6.72
78.3°C
24 x 0.125 gal.

Safety and Handling

DOTInformation : DOT Class: 3, Packaging: II

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit