Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanol, 200 proof (100%), USP, Decon™ Labs
Click to view available options
Quantity:
0.125 gal.
5 gal.
4 x 1 gal.
55 gal.
24 x 0.125 gal.
Packaging:
Cubitainer with Spigot
Glass Bottle
Metal Container
Plastic Bottle
Poly Bottle
Poly Bottle (Closure can be fitted with spigot)
Poly Drum
Description
Used for research, healthcare, industrial, or production applications as a cleaner, solvent or reagent.
Specifications
Melting Point | -114°C |
CAS Min % | 99.9 |
CAS Max % | 100.0 |
Density | 6.62 |
Boiling Point | 78.3°C |
Quantity | 5 gal. |
Packaging | Poly Bottle |
Physical Form | Liquid |
UN Number | UN1170 |
Grade | BP/EP/FCC/JP/NF/USP |
Show More |
Safety and Handling
DOTInformation | DOT Class: 3, Packaging II |
Recommended Storage | Do not store Ethanol at temperature above 120°F |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction