Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK

Manufacturer:  Alfa Aesar™ 36642AP

Quantity 500mL

 View more versions of this product

Catalog No. AA36642AP


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

  • As a gasoline additive/substitute; octane booster
  • Very mobile liquid solvent
  • Used in lotions, tonics and perfumes
Description & Specifications

Chemical Identifiers

64-17-5
CH3CH2OH
MFCD00003568
1718733
Ethyl alcohol
C2H6O
46.08
UN1993
14,3760
Ethanol, Anhydrous, denatured

Specifications

64-17-5
85.8%
CH3CH2OH
MFCD00003568
1718733
Ethyl alcohol
13.3% methanol, 0.9% MIBK
Like alcohol
11° (52°F)
Colorless
500mL
Ethanol, Anhydrous, denatured
Liquid
C2H6O
46.08
UN1993
14,3760
Miscible in water and most organic solvents
0.811g/mL
1.3610
80°C
−100°C
Hygroscopic
85.8%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.

GHS P Statement
P210-P260-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : KQ6300000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit