Learn More
Ethanol, CDA 19-1, 200 Proof, PHARMCO-AAPER™
Appplications include production of cleaning products, paint thinners, and brake fluid
Manufacturer:  Pharmco Products 1230191200DM55M
Packaging Metal drum
Quantity 55 gal.
 View more versions of this product
Catalog No. 16-100-775
Due to product restrictions, please to purchase or view availability for this product.
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-5,108-10-1,8008-20-6
C2H6O
Completely Denatured Alcohol
Liquid
46.07
Specifications
64-17-5,108-10-1,8008-20-6
C2H6O
Completely Denatured Alcohol
Metal drum
Carbon oxides produce hazardous products under combustion conditions
Clear colorless
Liquid
46.07
6.7 lb./gal.
Stable under recommended storage conditions.
Liquid
55 gal.
Safety and Handling
GHS Signal Word: Danger