Learn More
Ethanol, CDA 19-5, 200 Proof, PHARMCO-AAPER™
Appplications include production of cleaning products, paint thinners, and brake fluid
Manufacturer:  Pharmco Products 1230195200PL05M
Packaging Metal pail
Quantity 5 gal.
 View more versions of this product
Catalog No. 16-100-785
Due to product restrictions, please to purchase or view availability for this product.
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-5,108-10-1,64742-89-8
C2H6O
Completely Denatured Alcohol
Liquid
46.07
Specifications
64-17-5,108-10-1,64742-89-8
C2H6O
Completely Denatured Alcohol
Metal pail
Carbon oxides produce hazardous products under combustion conditions
Clear colorless
Liquid
46.07
6.79 lb./gal.
Stable under recommended storage conditions.
Liquid
5 gal.
Safety and Handling
GHS Signal Word: Danger