Learn More
Ethanol, CDA 19-6, 200 Proof, PHARMCO-AAPER™
Appplications include production of cleaning products, paint thinners, and brake fluid
Manufacturer:  Pharmco Products 1230196200CSGF
Packaging Fluorinated Poly Bottle
Quantity 1 gal.
 View more versions of this product
Catalog No. 16-100-794
Due to product restrictions, please to purchase or view availability for this product.
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-5,108-10-1,142-82-5
C2H6O
Completely Denatured Alcohol
Liquid
46.07
Specifications
64-17-5,108-10-1,142-82-5
C2H6O
Completely Denatured Alcohol
Fluorinated Poly Bottle
Carbon oxides produce hazardous products under combustion conditions
Clear colorless
Liquid
46.07
6.79 lb./gal.
Stable under recommended storage conditions.
Liquid
1 gal.
Safety and Handling
GHS Signal Word: Danger