Learn More
Ethanol-d{6}, 99.5%(Isotopic), Thermo Scientific Chemicals
Description
Persimmon, the fruit treated with ethanol-d_6, thioethers 9d and 9e were obtained, indicating that the C_2 unit originated from acetaldehyde which generated by oxidation of ethanol added externally.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Color | Colorless |
Quantity | 5 g |
Density | 0.91 g/mL |
Odor | Alcohol-like |
Sensitivity | Hygroscopic |
Boiling Point | 79°C |
Linear Formula | CD3CD2OD |
UN Number | UN1170 |
Solubility Information | Soluble in water. |
Show More |
Safety and Handling
Hazard Category | H224 |
Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS |
EINECSNumber | 216-162-2 |
TSCA | No |
Recommended Storage | Ambient |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.