Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
Description
Ethanol, anhydrous, denatured is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 90°C |
Color | Colorless |
Assay Percent Range | 90% |
UN Number | 1987 |
Appearance | Colorless liquid |
Beilstein | 1718733 |
Solubility Information | Miscible with water and most organic solvents. |
Formula Weight | 46.08 |
Grade | Spectrophotometric |
Concentration or Composition (by Analyte or Components) | Ethyl alcohol: 90.0%; Methanol: 5.0%; 2-Propanol: 5.0% |
Show More |
Safety and Handling
Hazard Category | H225-H302+H332-H319-H336-H371 |
Hazard Statement | Keep container tightly closed and in a well-ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P309+P311-P312-P330-P370+P378q-P501c |
DOTInformation | Hazard Class: 3, Packing Group: II |
EINECSNumber | 200-578-6 |
RTECSNumber | KQ6300000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
For Manufacturing and Laboratory Use. Not for beverage use
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction