Learn More
Ethanol, Prop Solv III-1, 200 Proof, PHARMCO-AAPER™
Manufacturer:  Pharmco Products 125PSIII1200DM55M
Packaging Metal drum
Quantity 55 gal.
 View more versions of this product
Catalog No. 16-101-237
Due to product restrictions, please to purchase or view availability for this product.
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-5,67-56-1,108-10-1,141-78-6,64742-89-8
C2H6O
Denatured Ethanol
Liquid
46.07
Specifications
64-17-5,67-56-1,108-10-1,141-78-6,64742-89-8
C2H6O
Denatured Ethanol
Stable under recommended storage conditions.
Liquid
55 gal.
Liquid
46.07
Metal drum
Carbon oxides produce hazardous products under combustion conditions
Clear colorless
Safety and Handling
GHS Signal Word: Danger