Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, SDA 23A, 190 Proof, PHARMCO-AAPER™

Applications include pharmaceutical and biotech processes, cosmetic, food, flavor, and fragrance industries

Manufacturer:  Pharmco Products 12323A190CSGF

Packaging Fluorinated Poly Bottle
Quantity 1 gal.

 View more versions of this product

Catalog No. 16-100-290


  Check Availability
Add to cart

Alert:

Minimum Order Quantity: 5


Specifications

Description

Order Info

Minimum Order Quantity: 5

Description & Specifications

Chemical Identifiers

64-17-15,7732-18-5,67-64-1
C2H6O
Denatured Alcohol 23A
Liquid, Invisible Vapor
46.07

Specifications

64-17-15,7732-18-5,67-64-1
C2H6O
Denatured Alcohol 23A
Sweet, alcohol-like
Stable under recommended storage conditions.
Liquid
1 gal.
Liquid, Invisible Vapor
46.07
0.785g/mL at 25°C (77°F)
Fluorinated Poly Bottle
Carbon oxides produce hazardous products under combustion conditions
Colorless

Safety and Handling

GHS Signal Word: Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit