Ethanol, SDA 28A-1, 190 Proof, PHARMCO-AAPER™

As a fuel: Automotive, airplane, rocket, jet, heating and other supplementary and misc fuels

Manufacturer: Pharmco Products 12328A1190CSGF

Packaging Fluorinated Poly Bottle
Quantity 1 gal.

 View more versions of this product

Catalog No. 16-100-584

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-17-5,7732-18-5,142-82-5
    Physical Form Liquid, Invisible Vapor
    Molecular Formula C2H6O
    Formula Weight 46.07
    Synonym Denatured Alcohol 28A-1

    Specifications

    Physical Form Liquid, Invisible Vapor
    Color Colorless
    Formula Weight 46.07
    Quantity 1 gal.
    Decomposition Information Carbon oxides produce hazardous products under combustion conditions
    Physical State Liquid
    Density 6.61 lb./gal. at 15.56°C (60°F)
    Odor Sweet, alcohol-like
    Packaging Fluorinated Poly Bottle
    Stability Stable under recommended storage conditions.
    Synonym Denatured Alcohol 28A-1
    Health Hazard 1 GHS Signal Word: Danger
    Molecular Formula C2H6O
    CAS 64-17-5,7732-18-5,142-82-5

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

    GHS Signal Word: Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit