Ethanol, SDA 38B-09, 190 Proof, PHARMCO-AAPER™

Manufacturer: Pharmco Products 12338B09190PL05M

Packaging Metal pail
Quantity 5 gal.

 View more versions of this product

Catalog No. 16-100-681

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-17-5,7732-18-5,97-53-0,8007-80-5
    Physical Form Liquid, Invisible Vapor
    Molecular Formula C2H6O
    Formula Weight 46.07
    Synonym Denatured Alcohol 38B-09

    Specifications

    Physical Form Liquid, Invisible Vapor
    Formula Weight 46.07
    Color Colorless
    Quantity 5 gal.
    Decomposition Information Carbon oxides produce hazardous products under combustion conditions
    Physical State Liquid
    Density 6.62 lb./gal. at 15.56°C (60°F)
    CAS 8007-80-5,7732-18-5,64-17-5,97-53-0
    Packaging Metal pail
    Stability Stable under recommended storage conditions.
    Molecular Formula C2H6O
    Synonym Denatured Alcohol 38B-09
    Health Hazard 1 GHS Signal Word: Danger
    Odor Characteristic

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

    GHS Signal Word: Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit