Learn More
Ethanol, SDA 38B-33, 200 Proof, PHARMCO-AAPER™
Manufacturer:  Pharmco Products 1238B33200PL05M
Packaging Metal pail
Quantity 5 gal.
 View more versions of this product
Catalog No. 16-100-357
  Check Availability
Add to cart
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-15,89-83-8,8006-90-4,119-36-8,89-78-1,470-82-6
C2H6O
Ethanol denatured with Thymol Crystals, Menthol, Methyl Salicylate
Liquid, Invisible Vapor
46.07
Specifications
64-17-15,89-83-8,8006-90-4,119-36-8,89-78-1,470-82-6
C2H6O
Ethanol denatured with Thymol Crystals, Menthol, Methyl Salicylate
Sweet, alcohol-like
Stable under recommended storage conditions.
Liquid
5 gal.
Liquid, Invisible Vapor
46.07
0.796g/mL at 25C (77F)
Metal pail
Carbon oxides produce hazardous products under combustion conditions
Colorless
Safety and Handling
GHS Signal Word: Danger