Ethanol, SDA 38B-58, 200 Proof, PHARMCO-AAPER™

Manufacturer: Pharmco Products 12338B58200PL05

Packaging Poly pail
Quantity 5 gal.

 View more versions of this product

Catalog No. 16-100-416

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-17-15,97-53-0,470-82-6
    Physical Form Liquid, Invisible Vapor
    Molecular Formula C2H6O
    Formula Weight 46.07
    Synonym Denatured Alcohol 38B-58

    Specifications

    Physical Form Liquid, Invisible Vapor
    Color Colorless
    Formula Weight 46.07
    Quantity 5 gal.
    Decomposition Information Carbon oxides produce hazardous products under combustion conditions
    Physical State Liquid
    Density 6.62 lb./gal. at 15.56°C (60°F)
    CAS 64-17-15,470-82-6,97-53-0
    Odor Characteristic
    Packaging Poly pail
    Stability Stable under recommended storage conditions.
    Molecular Formula C2H6O
    Synonym Denatured Alcohol 38B-58
    Health Hazard 1 GHS Signal Word: Danger

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

    GHS Signal Word: Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit