Learn More
Ethanol, SDA 38B-83, 190 Proof, PHARMCO-AAPER™
Manufacturer:  Pharmco Products 12338B83190PL05M
Packaging Metal pail
Quantity 5 gal.
 View more versions of this product
Catalog No. 16-100-499
  Check Availability
Add to cart
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-15,7732-18-5,8007-75-8,118-55-8,8000-28-0
C2H6O
Denatured Alcohol 38B-83
Liquid, Invisible Vapor
46.07
Specifications
64-17-15,7732-18-5,8007-75-8,118-55-8,8000-28-0
C2H6O
Denatured Alcohol 38B-83
Characteristic
Stable under recommended storage conditions.
Liquid
5 gal.
Liquid, Invisible Vapor
46.07
6.622 lb./gal. at 15.56C (60F)
Metal pail
Carbon oxides produce hazardous products under combustion conditions
Colorless
Safety and Handling
GHS Signal Word: Danger