Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, SDA 4, 200 PROOF, PHARMCO-AAPER™

Tobacco sprays and flavors, product development and pilot plant uses (own use only)

Manufacturer:  Pharmco Products 123004200CS4L

Packaging Glass bottle
Quantity 4L

 View more versions of this product

Catalog No. 16-100-273


  Check Availability
Add to cart

Alert:

Minimum Order Quantity: 5


Specifications

Description

Order Info

Minimum Order Quantity: 5

Description & Specifications

Chemical Identifiers

64-17-15,65-30-5,7220-79-3
C2H6O
Denatured Alcohol (SDA) 4, 200
Liquid
46.07

Specifications

64-17-15,65-30-5,7220-79-3
C2H6O
Denatured Alcohol (SDA) 4, 200
Characteristic to ethanol
Stable under recommended storage conditions.
Liquid
4L
Liquid
46.07
6.823 lb./gal. at 15.56°C (60°F)
Glass bottle
Carbon oxides produce hazardous products under combustion conditions
Blue

Safety and Handling

GHS Signal Word: Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit