Ethanol, SIS 642, 200 Proof, PHARMCO-AAPER™

Applications include Industrial and manufacturing processes

Manufacturer: Pharmco Products 125642200DM55

Packaging Poly drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-100-880

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 64-17-5,67-56-1,109-60-4,108-10-1
    Physical Form Liquid
    Molecular Formula C2H6O
    Formula Weight 46.07
    Synonym Denatured Ethanol

    Specifications

    Physical Form Liquid
    Formula Weight 46.07
    Color Clear colorless
    Quantity 55 gal.
    Decomposition Information Carbon oxides produce hazardous products under combustion conditions
    Physical State Liquid
    CAS 108-10-1,64-17-5,67-56-1,109-60-4
    Packaging Poly drum
    Stability Stable under recommended storage conditions.
    Synonym Denatured Ethanol
    Molecular Formula C2H6O
    Health Hazard 1 GHS Signal Word: Danger

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

    GHS Signal Word: Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit