Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, Solvent KK008, 190 Proof, PHARMCO-AAPER™

Applications include Industrial and manufacturing processes

Manufacturer:  Pharmco Products 126KK8190DM55

Packaging Poly drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-108


 

Due to product restrictions, please to purchase or view availability for this product.

Alert:

Minimum Order Quantity: 5


Specifications

Description

Order Info

Minimum Order Quantity: 5

Description & Specifications

Chemical Identifiers

64-17-5,7732-18-5,67-56-1,108-10-1
C2H6O
Denatured Ethanol
Liquid
46.07

Specifications

64-17-5,7732-18-5,67-56-1,108-10-1
C2H6O
Denatured Ethanol
Stable under recommended storage conditions.
Liquid
55 gal.
Liquid
46.07
Poly drum
Carbon oxides produce hazardous products under combustion conditions
Clear colorless

Safety and Handling

GHS Signal Word: Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit