Learn More
Ethanol, TR-590, 200 Proof, PHARMCO-AAPER™
Industrial solvent use
Manufacturer:  Pharmco Products 125590200PL05M
Packaging Metal pail
Quantity 5 gal.
 View more versions of this product
Catalog No. 16-101-307
Due to product restrictions, please to purchase or view availability for this product.
Alert:
Minimum Order Quantity: 5
Specifications
Description
Order Info
Minimum Order Quantity: 5
Description & Specifications
Chemical Identifiers
64-17-5,141-78-6,67-56-1,108-10-1
C2H6O
Denatured Ethanol
Liquid
46.07
Specifications
64-17-5,141-78-6,67-56-1,108-10-1
C2H6O
Denatured Ethanol
Stable under recommended storage conditions.
Liquid
5 gal.
Liquid
46.07
Metal pail
Carbon oxides produce hazardous products under combustion conditions
Clear colorless
Safety and Handling
GHS Signal Word: Danger