Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethanolamine, 45% v/v Reagent 1, Reagents
Ethanolamine, 45% v/v Reagent 1, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CE0019004A
Quantity | 4 L |
---|
Chemical Identifiers
7732-18-5, 141-43-5, 7647-01-0 | |
18.02 | |
2-Aminoethanol, Monoethanolamine | |
O |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water |
Specifications
Colorless to Light Yellow | |
H2O | |
XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
water | |
45% | |
Poly Bottle | |
4 L | |
Liquid |
7732-18-5, 141-43-5, 7647-01-0 | |
2-Aminoethanol, Monoethanolamine | |
O | |
18.02 | |
Reagent | |
1.0198 | |
Non-sterile | |
Ethanolamine, 45% v/v Reagent 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction