Alfa Aesar™ Ethanolamine, 99%, H2O 0.5% max

Manufacturer: Alfa Aesar™ 22793A1

Quantity 1kg

 View more versions of this product

Catalog No. AA22793A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141-43-5
    Formula Weight 61.08
    MDL Number MFCD00008183
    UN Number UN2491
    Beilstein 505944
    Merck Index 14,3727
    Chemical Name or Material Ethanolamine

    Specifications

    Formula Weight 61.08
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 505944
    Health Hazard 1 Danger
    Refractive Index 1.4540
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.
    Boiling Point 169° to 170°C
    Quantity 1kg
    Flash Point 93° (199°F)
    Physical Form Liquid
    UN Number UN2491
    Density 1.012g/mL
    Sensitivity Air Sensitive & Hygroscopic
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE
    TSCA TSCA
    MDL Number MFCD00008183
    CAS 141-43-5
    Merck Index 14,3727
    Melting Point 10°C
    Odor Ammonia-like
    Concentration H2O 0.5% max.
    RTECS Number KJ5775000
    Chemical Name or Material Ethanolamine
    Assay 99%

    GHS H Statement
    H314-H318-H302-H312-H332-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit