Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, ACS, 99+%, Alfa Aesar™

Click to view available options
Quantity:
500mL
4 x 500mL
141-43-5
C2H7NO
61.084
MFCD00008183
HZAXFHJVJLSVMW-UHFFFAOYSA-N
ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine
700
CHEBI:16000
2-aminoethanol
C(CO)N
141-43-5
C2H7NO
61.084
MFCD00008183
HZAXFHJVJLSVMW-UHFFFAOYSA-N
ethanolamine,monoethanolamine,aminoethanol,2-hydroxyethylamine,colamine,glycinol,olamine,2-amino-1-ethanol,ethanol, 2-amino,ethylolamine
700
CHEBI:16000
2-aminoethanol
C(CO)N
Density 1.012g/mL
Quantity 500mL
Assay Percent Range >99+%
UN Number UN2491
Beilstein 505944
Merck Index 14,3727
Formula Weight 61.08
Physical Form Liquid
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Ethanolamine, For analysis ACS
Show More Show Less

GHS H Statement
H314-H302-H312-H332-H335-H227

GHS P Statement
P210-P260-P261-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ETHANOLAMINE

RTECSNumber : KJ5775000

TSCA : TSCA