Learn More
ETHE1 Mouse anti-Human, Mouse, Rat, Alexa Fluor™ 350, Clone: 1621, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243632AF350
Catalog No. NP243632A30
Add to cart
Description
Description
ETHE1 Monoclonal antibody specifically detects ETHE1 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
ETHE1
1621
Western Blot, Immunocytochemistry/Immunofluorescence
D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ETHE1
Full length human ETHE1 Recombinant protein.
0.1 mL
Primary
23474