Learn More
ETHE1 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: 1621, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243632IR
Catalog No. NBP243632IR
Add to cart
Description
Description
ETHE1 Monoclonal antibody specifically detects ETHE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
ETHE1
1621
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
ETHE1
Full length human ETHE1 Recombinant protein.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
D83198, EC 3.1.2.6, ethylmalonic encephalopathy 1, Ethylmalonic encephalopathy protein 1, Hepatoma subtracted clone one protein, HSCOEC 3.-, protein ETHE1, mitochondrial, YF13H12
Mouse
IgG2b
0.1 ml
Primary
23474